siRNA Details
| Gene Symbol: Tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108670.1 | NP_001102140.1 | 7 | 966 | ||
| XM_006238417.2 | XP_006238479.1 | 7 | 848 | |||
| XM_006238418.2 | XP_006238480.1 | 7 | 848 | |||
| XM_006238419.2 | XP_006238481.1 | 6 | 743 | |||

Tm2d1
Genomic Map