siRNA Details
| Gene Symbol: Itgb3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013213.1 | NP_001013231.1 | 7, 8 | 807 | ||
| XM_008763882.1 | XP_008762104.1 | 5, 6 | 624 | |||
| XM_008763883.1 | XP_008762105.1 | 8, 9 | 549 | |||
| XM_008763884.1 | XP_008762106.1 | 7, 8 | 503 | |||
| GenBank mRNA | BC086363.1 | - | 7, 8 | 807 | 7304501 | |

Itgb3bp
Genomic Map