siRNA Details
| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108671.1 | NP_001102141.1 | 7 | 1554 | ||
| XM_008763887.1 | XP_008762109.1 | 6 | 2404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ror1| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108671.1 | NP_001102141.1 | 7 | 1554 | ||
| XM_008763887.1 | XP_008762109.1 | 6 | 2404 | |||