siRNA Details
| Gene Symbol: Tctex1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135030.1 | NP_001128502.1 | 3, 4 | 419 | ||
| XM_006238459.2 | XP_006238521.1 | 3, 4 | 1175 | |||
| XM_008763888.1 | XP_008762110.1 | 2, 3 | 246 | |||
| XM_008763889.1 | XP_008762111.1 | 3, 4 | 3092 | |||
| XM_008763890.1 | XP_008762112.1 | 2, 3 | 953 | |||
| XM_008763891.1 | XP_008762113.1 | 2, 3 | 396 | |||

Tctex1d1
Genomic Map