siRNA Details
| Gene Symbol: Bend5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108672.1 | NP_001102142.1 | 5 | 1136 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bend5| Gene Symbol: Bend5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108672.1 | NP_001102142.1 | 5 | 1136 | ||