siRNA Details
| Gene Symbol: Pomgnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007747.1 | NP_001007748.1 | 9 | 1002 | ||
| XM_006238683.2 | XP_006238745.1 | 8 | 1109 | |||
| GenBank mRNA | BC083641.1 | - | 9 | 1002 | 7134894 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pomgnt1| Gene Symbol: Pomgnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007747.1 | NP_001007748.1 | 9 | 1002 | ||
| XM_006238683.2 | XP_006238745.1 | 8 | 1109 | |||
| GenBank mRNA | BC083641.1 | - | 9 | 1002 | 7134894 | |