siRNA Details
| Gene Symbol: RGD1305347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108674.1 | NP_001102144.1 | 3 | 694 | ||
| XM_006238692.2 | XP_006238754.1 | 3 | 590 | |||
| XM_006238693.2 | XP_006238755.1 | 4 | 985 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305347| Gene Symbol: RGD1305347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108674.1 | NP_001102144.1 | 3 | 694 | ||
| XM_006238692.2 | XP_006238754.1 | 3 | 590 | |||
| XM_006238693.2 | XP_006238755.1 | 4 | 985 | |||