siRNA Details
| Gene Symbol: Ccdc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038992.1 | NP_001034081.1 | 2, 3 | 253 | ||
| NM_001038994.2 | NP_001034083.1 | 2, 3 | 353 | |||
| GenBank mRNA | BC099226.1 | - | 2, 3 | 253 | 7388643 | |
| BF559487.1 | - | 2, 3 | 359 | |||

Ccdc23
Genomic Map