siRNA Details
| Gene Symbol: Col9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108675.1 | NP_001102145.1 | 32 | 2758 | ||
| XM_008764033.1 | XP_008762255.1 | 31 | 2745 | |||
| GenBank mRNA | FQ211982.1 | - | 34 | 2546 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col9a2| Gene Symbol: Col9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108675.1 | NP_001102145.1 | 32 | 2758 | ||
| XM_008764033.1 | XP_008762255.1 | 31 | 2745 | |||
| GenBank mRNA | FQ211982.1 | - | 34 | 2546 | ||