siRNA Details
| Gene Symbol: Col9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108675.1 | NP_001102145.1 | 27, 28 | 1781 | ||
| XM_008764033.1 | XP_008762255.1 | 27, 28 | 1485 | |||
| GenBank mRNA | FQ211982.1 | - | 29, 30 | 1569 | ||

Col9a2
Genomic Map