siRNA Details
| Gene Symbol: LOC100909856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764120.1 | XP_008762342.1 | 4 | 914 | ||
| Gene Symbol: Mtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108677.1 | NP_001102147.1 | 4 | 888 | ||
| XM_006238883.2 | XP_006238945.1 | 4 | 1487 | |||
| XM_006238884.2 | XP_006238946.1 | 4 | 914 | |||

LOC100909856
Genomic Map