siRNA Details
| Gene Symbol: Zmym6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108681.2 | NP_001102151.1 | 15 | 2675 | ||
| XM_006238892.2 | XP_006238954.1 | 14 | 3506 | |||
| XM_006238894.2 | XP_006238956.1 | 15 | 2553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmym6| Gene Symbol: Zmym6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108681.2 | NP_001102151.1 | 15 | 2675 | ||
| XM_006238892.2 | XP_006238954.1 | 14 | 3506 | |||
| XM_006238894.2 | XP_006238956.1 | 15 | 2553 | |||