siRNA Details
| Gene Symbol: Sync | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108683.1 | NP_001102153.1 | 2 | 368 | ||
| XM_006238943.2 | XP_006239005.1 | 2 | 438 | |||
| XM_006238944.2 | XP_006239006.1 | 2 | 437 | |||
| XM_006238945.2 | XP_006239007.1 | 2 | 438 | |||

Sync
Genomic Map