siRNA Details
| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270959.1 | NP_001257888.1 | 4 | 748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpn2| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270959.1 | NP_001257888.1 | 4 | 748 | ||