siRNA Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134956.1 | NP_001128428.1 | 5 | 2222 | ||
| XM_008764151.1 | XP_008762373.1 | 3 | 1948 | |||
| XM_008764152.1 | XP_008762374.1 | 3 | 1843 | |||
| XM_008764153.1 | XP_008762375.1 | 3 | 1986 | |||
| XM_008764154.1 | XP_008762376.1 | 3 | 1962 | |||

Ahdc1
Genomic Map