siRNA Details
| Gene Symbol: Smpdl3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025737.1 | NP_001020908.1 | 7 | 1078 | ||
| GenBank mRNA | BC097983.1 | - | 7 | 1078 | 7443444 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smpdl3b| Gene Symbol: Smpdl3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025737.1 | NP_001020908.1 | 7 | 1078 | ||
| GenBank mRNA | BC097983.1 | - | 7 | 1078 | 7443444 | |