siRNA Details
Gene Symbol: Ifnlr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191868.1 | NP_001178797.1 | 2, 3 | 182 | ||
XM_006239219.2 | XP_006239281.1 | 1, 2 | 1108 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifnlr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191868.1 | NP_001178797.1 | 2, 3 | 182 | ||
XM_006239219.2 | XP_006239281.1 | 1, 2 | 1108 |