siRNA Details
| Gene Symbol: Srsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025738.1 | NP_001020909.1 | 4 | 497 | ||
| XM_006239220.2 | XP_006239282.1 | 4 | 540 | |||
| XM_006239221.2 | XP_006239283.1 | 4 | 540 | |||
| XM_006239222.2 | XP_006239284.1 | 4 | 538 | |||
| XM_006239223.2 | XP_006239285.1 | 4 | 539 | |||
| GenBank mRNA | BC098831.1 | - | 4 | 497 | 7378554 | |
| FQ227288.1 | - | 4 | 518 | |||

Srsf10
Genomic Map