siRNA Details
| Gene Symbol: C1qc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008524.1 | NP_001008524.1 | 3 | 636 | ||
| XM_006239225.1 | XP_006239287.1 | 3 | 916 | |||
| GenBank mRNA | BC086409.1 | - | 3 | 636 | 7306579 | |
| FQ218827.1 | - | 3 | 660 | |||
| FQ219610.1 | - | 3 | 688 | |||
| FQ220269.1 | - | 3 | 688 | |||
| FQ228978.1 | - | 3 | 920 | |||

C1qc
Genomic Map