siRNA Details
| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173556.1 | NP_001167027.1 | 4 | 329 | ||
| GenBank mRNA | FM054041.1 | - | 4 | 329 | ||
| FQ221358.1 | - | 4 | 257 | |||
| FQ228668.1 | - | 4 | 331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Minos1| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173556.1 | NP_001167027.1 | 4 | 329 | ||
| GenBank mRNA | FM054041.1 | - | 4 | 329 | ||
| FQ221358.1 | - | 4 | 257 | |||
| FQ228668.1 | - | 4 | 331 | |||