siRNA Details
| Gene Symbol: Fbxo42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108691.1 | NP_001102161.1 | 2 | 371 | ||
| XM_006239253.2 | XP_006239315.1 | 2 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo42| Gene Symbol: Fbxo42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108691.1 | NP_001102161.1 | 2 | 371 | ||
| XM_006239253.2 | XP_006239315.1 | 2 | 371 | |||