siRNA Details
| Gene Symbol: Ctrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077649.2 | NP_001071117.1 | 7 | 737 | ||
| GenBank mRNA | S80379.1 | - | 7 | 738 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctrc| Gene Symbol: Ctrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077649.2 | NP_001071117.1 | 7 | 737 | ||
| GenBank mRNA | S80379.1 | - | 7 | 738 | ||