siRNA Details
| Gene Symbol: Zbtb48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013216.2 | NP_001013234.2 | 7 | 1370 | ||
| XM_006239469.2 | XP_006239531.1 | 7 | 1379 | |||
| XM_008764367.1 | XP_008762589.1 | 7 | 1408 | |||
| GenBank mRNA | BC091214.1 | - | 6 | 1969 | 7383542 | |
| FQ211844.1 | - | 7 | 1397 | |||

Zbtb48
Genomic Map