siRNA Details
| Gene Symbol: RGD1308923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108698.1 | NP_001102168.1 | 3 | 463 | ||
| XM_006239585.2 | XP_006239647.1 | 3 | 1144 | |||
| GenBank mRNA | FQ234838.1 | - | 3 | 229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1308923| Gene Symbol: RGD1308923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108698.1 | NP_001102168.1 | 3 | 463 | ||
| XM_006239585.2 | XP_006239647.1 | 3 | 1144 | |||
| GenBank mRNA | FQ234838.1 | - | 3 | 229 | ||