siRNA Details
| Gene Symbol: Gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108700.1 | NP_001102170.1 | 6, 7 | 692 | ||
| XM_006239636.2 | XP_006239698.1 | 6, 7 | 968 | |||
| XM_008764424.1 | XP_008762646.1 | 7, 8 | 809 | |||
| GenBank mRNA | FQ225505.1 | - | 6, 7 | 699 | ||

Gpatch11
Genomic Map