siRNA Details
| Gene Symbol: Srsf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039035.2 | NP_001034124.2 | 2 | 197 | ||
| XM_006239642.1 | XP_006239704.1 | 2 | 237 | |||
| XM_006239643.1 | XP_006239705.1 | 2 | 237 | |||
| XM_006239644.1 | XP_006239706.1 | 2 | 237 | |||
| XM_006239645.1 | XP_006239707.1 | 2 | 237 | |||
| XM_006239646.1 | XP_006239708.1 | 2 | 237 | |||
| GenBank mRNA | BC099175.1 | - | 2 | 187 | 7382701 | |
| FM084661.1 | - | 2 | 197 | |||
| FM085512.1 | - | 2 | 191 | |||
| FQ219847.1 | - | 2 | 203 | |||
| FQ229134.1 | - | 2 | 204 | |||

Srsf7
Genomic Map