siRNA Details
| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271305.1 | NP_001258234.1 | 13, 14 | 1449 | ||
| XM_008764482.1 | XP_008762704.1 | 12, 13 | 1362 | |||
| GenBank mRNA | AY390384.1 | - | 13, 14 | 1300 | ||

Ppp1r21
Genomic Map