siRNA Details
| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009712.1 | NP_001009712.1 | 6 | 845 | ||
| GenBank mRNA | BC088468.1 | - | 6 | 845 | 7127993 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf4| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009712.1 | NP_001009712.1 | 6 | 845 | ||
| GenBank mRNA | BC088468.1 | - | 6 | 845 | 7127993 | |