siRNA Details
| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009712.1 | NP_001009712.1 | 1, 2 | 285 | ||
| GenBank mRNA | BC088468.1 | - | 1, 2 | 285 | 7127993 | |
| Gene Symbol: RGD1565766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239851.2 | XP_006239913.1 | 1 | 211 | ||

Yipf4
Genomic Map