siRNA Details
| Gene Symbol: Capn13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025133.1 | NP_001020304.1 | 11 | 1187 | ||
| XM_006239777.2 | XP_006239839.1 | 12 | 1345 | |||
| XM_006239778.2 | XP_006239840.1 | 11 | 1298 | |||
| XM_006239779.2 | XP_006239841.1 | 12 | 1341 | |||
| GenBank mRNA | BC091250.1 | - | 11 | 1187 | 7368359 | |

Capn13
Genomic Map