siRNA Details
| Gene Symbol: Rbks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108703.1 | NP_001102173.1 | 4 | 368 | ||
| XM_006239805.2 | XP_006239867.1 | 4 | 414 | |||
| XM_006239806.2 | XP_006239868.1 | 4 | 414 | |||
| XM_008764520.1 | XP_008762742.1 | 4 | 862 | |||
| XM_008764521.1 | XP_008762743.1 | 4 | 414 | |||
| GenBank mRNA | FQ209797.1 | - | 4 | 350 | ||

Rbks
Genomic Map