siRNA Details
| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013217.1 | NP_001013235.1 | 5 | 818 | ||
| GenBank mRNA | BC083706.1 | - | 5 | 818 | 7190356 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim54| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013217.1 | NP_001013235.1 | 5 | 818 | ||
| GenBank mRNA | BC083706.1 | - | 5 | 818 | 7190356 | |