siRNA Details
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014195.2 | NP_001014217.2 | 2 | 304 | ||
GenBank mRNA | BC129115.1 | - | 2 | 304 | 7112909 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014195.2 | NP_001014217.2 | 2 | 304 | ||
GenBank mRNA | BC129115.1 | - | 2 | 304 | 7112909 |