siRNA Details
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014195.2 | NP_001014217.2 | 10 | 1268 | ||
GenBank mRNA | BC085114.1 | - | 7 | 699 | 7106107 | |
BC129115.1 | - | 10 | 1268 | 7112909 |
