siRNA Details
| Gene Symbol: Agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135899.1 | NP_001129371.1 | 1 | 246 | ||
| GenBank mRNA | BC127545.1 | - | 1 | 246 | 7321962 | |
| FQ219104.1 | - | 1 | 290 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agmo| Gene Symbol: Agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135899.1 | NP_001129371.1 | 1 | 246 | ||
| GenBank mRNA | BC127545.1 | - | 1 | 246 | 7321962 | |
| FQ219104.1 | - | 1 | 290 | |||