siRNA Details
| Gene Symbol: Hectd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240086.1 | XP_006240148.1 | 18 | 2912 | ||
| XM_006240087.1 | XP_006240149.1 | 18 | 2912 | |||
| XM_006240088.1 | XP_006240150.1 | 18 | 2912 | |||
| XM_006240089.1 | XP_006240151.1 | 18 | 2912 | |||

Hectd1
Genomic Map