siRNA Details
| Gene Symbol: Heatr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240091.1 | XP_006240153.1 | 31 | 4938 | ||
| XM_006240095.1 | XP_006240157.1 | 30 | 4911 | |||
| XM_006240096.1 | XP_006240158.1 | 30 | 4908 | |||
| XM_008764667.1 | XP_008762889.1 | 31 | 4938 | |||
| XM_008764668.1 | XP_008762890.1 | 31 | 4935 | |||

Heatr5a
Genomic Map