siRNA Details
| Gene Symbol: Snx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108711.1 | NP_001102181.1 | 2, 3 | 98 | ||
| GenBank mRNA | BC168856.1 | - | 1, 2 | 59 | 7458822 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx6| Gene Symbol: Snx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108711.1 | NP_001102181.1 | 2, 3 | 98 | ||
| GenBank mRNA | BC168856.1 | - | 1, 2 | 59 | 7458822 | |