siRNA Details
| Gene Symbol: Pax9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039539.1 | NP_001034628.1 | 2 | 555 | ||
| GenBank mRNA | AB248957.1 | - | 2 | 555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax9| Gene Symbol: Pax9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039539.1 | NP_001034628.1 | 2 | 555 | ||
| GenBank mRNA | AB248957.1 | - | 2 | 555 | ||