siRNA Details
| Gene Symbol: Trmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108713.1 | NP_001102183.1 | 5 | 1448 | ||
| XM_006240213.2 | XP_006240275.1 | 4 | 1424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trmt5| Gene Symbol: Trmt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108713.1 | NP_001102183.1 | 5 | 1448 | ||
| XM_006240213.2 | XP_006240275.1 | 4 | 1424 | |||