siRNA Details
| Gene Symbol: Rdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012193.1 | NP_001012193.1 | 5 | 553 | ||
| GenBank mRNA | AY387097.1 | - | 5 | 721 | ||
| BC079276.1 | - | 5 | 553 | 7134077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh11| Gene Symbol: Rdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012193.1 | NP_001012193.1 | 5 | 553 | ||
| GenBank mRNA | AY387097.1 | - | 5 | 721 | ||
| BC079276.1 | - | 5 | 553 | 7134077 | ||