siRNA Details
| Gene Symbol: Galnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100863.1 | NP_001094333.1 | 9 | 1051 | ||
| XM_006240288.2 | XP_006240350.1 | 10 | 2182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt16| Gene Symbol: Galnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100863.1 | NP_001094333.1 | 9 | 1051 | ||
| XM_006240288.2 | XP_006240350.1 | 10 | 2182 | |||