siRNA Details
| Gene Symbol: Angel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108717.1 | NP_001102187.1 | 2, 3 | 697 | ||
| XM_006240392.2 | XP_006240454.1 | 3, 4 | 1229 | |||
| XM_008764815.1 | XP_008763037.1 | 2, 3 | 885 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Angel1| Gene Symbol: Angel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108717.1 | NP_001102187.1 | 2, 3 | 697 | ||
| XM_006240392.2 | XP_006240454.1 | 3, 4 | 1229 | |||
| XM_008764815.1 | XP_008763037.1 | 2, 3 | 885 | |||