siRNA Details
| Gene Symbol: Alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108718.1 | NP_001102188.1 | 3, 4 | 479 | ||
| XM_006240394.2 | XP_006240456.1 | 3, 4 | 485 | |||
| GenBank mRNA | FQ225419.1 | - | 1, 2 | 60 | ||
| FQ230972.1 | - | 1, 2 | 60 | |||

Alkbh1
Genomic Map