siRNA Details
| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764818.1 | XP_008763040.1 | 9 | 2453 | ||
| XM_008764819.1 | XP_008763041.1 | 10 | 2595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd7| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764818.1 | XP_008763040.1 | 9 | 2453 | ||
| XM_008764819.1 | XP_008763041.1 | 10 | 2595 | |||