siRNA Details
| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108720.1 | NP_001102190.1 | 2 | 805 | ||
| XM_008764818.1 | XP_008763040.1 | 2 | 840 | |||
| XM_008764819.1 | XP_008763041.1 | 3 | 982 | |||
| XM_008764820.1 | XP_008763042.1 | 2 | 841 | |||
| XM_008764821.1 | XP_008763043.1 | 2 | 841 | |||

Btbd7
Genomic Map