siRNA Details
| Gene Symbol: Serpina11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008776.2 | NP_001008776.1 | 5 | 1204 | ||
| NM_001166352.1 | NP_001159824.1 | 5 | 1190 | |||
| XM_006240481.2 | XP_006240543.1 | 6 | 1309 | |||
| GenBank mRNA | AY325135.1 | - | 5 | 1244 | ||
| BC111708.1 | - | 4 | 1102 | 7368690 | ||
| FQ211426.1 | - | 5 | 1162 | |||

Serpina11
Genomic Map