siRNA Details
| Gene Symbol: Mok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010965.1 | NP_001010965.1 | 2 | 240 | ||
| XM_006240589.2 | XP_006240651.2 | 2 | 400 | |||
| XM_006240590.2 | XP_006240652.2 | 2 | 400 | |||
| XM_006240592.2 | XP_006240654.2 | 2 | 400 | |||
| XM_006240593.2 | XP_006240655.2 | 2 | 400 | |||
| XM_008764944.1 | XP_008763166.1 | 2 | 400 | |||
| GenBank mRNA | BC079440.1 | - | 2 | 240 | 7115407 | |

Mok
Genomic Map