siRNA Details
| Gene Symbol: Inf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764994.1 | XP_008763216.1 | 8 | 2917 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inf2| Gene Symbol: Inf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764994.1 | XP_008763216.1 | 8 | 2917 | ||