siRNA Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126288.1 | NP_001119760.1 | 9, 10 | 1308 | ||
| GenBank mRNA | BC161906.1 | - | 9, 10 | 1216 | 8367484 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pld4| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126288.1 | NP_001119760.1 | 9, 10 | 1308 | ||
| GenBank mRNA | BC161906.1 | - | 9, 10 | 1216 | 8367484 | |